Learn More
Cayman Chemical THUNDRTotal p70 S6K TRFRE 4800
SDP
Catalog No. 502895617
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total p70 S6K in cell lysates.
Catalog No. 50-289-5617 Supplier Cayman Chemical Supplier No. 5002754800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total p70 S6K in cell lysates.