Learn More
Cayman Chemical THUNDRTotal RK12 TRFRET 4800
SDP
Catalog No. 502892625
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total ERK1/2 (both phosphorylated and non-phosphorylated ERK1/2) in cell lysates
Catalog No. 50-289-2625 Supplier Cayman Chemical Supplier No. 5002074800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total ERK1/2 (both phosphorylated and non-phosphorylated ERK1/2) in cell lysates