Learn More
Cayman Chemical THUNDRTotal RK12 TRFRET 96Wel
SDP
Catalog No. 502895435
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total ERK1/2 (both phosphorylated and non-phosphorylated ERK1/2) in cell lysates
Catalog No. 50-289-5435 Supplier Cayman Chemical Supplier No. 50020796 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total ERK1/2 (both phosphorylated and non-phosphorylated ERK1/2) in cell lysates