Learn More
Cayman Chemical THUNDRTotal RPS6 TRFRET C 2400
SDP
Catalog No. 502895262
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total Ribosomal Protein S6 (both phosphorylated and non-phosphorylated RPS6) in cell lysates.
Catalog No. 50-289-5262 Supplier Cayman Chemical Supplier No. 5002792400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total Ribosomal Protein S6 (both phosphorylated and non-phosphorylated RPS6) in cell lysates.