Learn More
Cayman Chemical THUNDRTotal RPS6 TRFRET C 96
SDP
Catalog No. 502893480
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total Ribosomal Protein S6 (both phosphorylated and non-phosphorylated RPS6) in cell lysates.
Catalog No. 50-289-3480 Supplier Cayman Chemical Supplier No. 50027996 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total Ribosomal Protein S6 (both phosphorylated and non-phosphorylated RPS6) in cell lysates.