Learn More
Cayman Chemical THUNDRTotal RPS6 TRFRET C 9600
SDP
Catalog No. 502894446
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total Ribosomal Protein S6 (both phosphorylated and non-phosphorylated RPS6) in cell lysates.
Catalog No. 50-289-4446 Supplier Cayman Chemical Supplier No. 5002799600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total Ribosomal Protein S6 (both phosphorylated and non-phosphorylated RPS6) in cell lysates.