Learn More
Cayman Chemical THUNDRTotal STAT5 TRFRET 2400
SDP
Catalog No. 502896037
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-6037 Supplier Cayman Chemical Supplier No. 5002162400 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates