Learn More
Cayman Chemical THUNDRTotal STAT5 TRFRET 4800
SDP
Catalog No. 502895923
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-5923 Supplier Cayman Chemical Supplier No. 5002164800 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates