Learn More
Cayman Chemical THUNDRTotal STAT5 TRFRET 9600
SDP
Catalog No. 502894563
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-4563 Supplier Cayman Chemical Supplier No. 5002169600 WELL
Add to Cart
Add to Cart
Request a Quote
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates