Learn More
Cayman Chemical THUNDRTotal STAT5 TRFRET 96Wel
SDP
Catalog No. 502895959
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-5959 Supplier Cayman Chemical Supplier No. 50021696 WELL
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT5 (both phosphorylated and non-phosphorylated) in cell lysates