Learn More
Enzo Life Sciences Thy-1 (human) : Fc (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522091C050
Encompass_Preferred
Alternative name: CD90. Concentration: 1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human Thy-1 (aa 20-130) is fused to the Fc portion of human IgG1. Long Term Storage: -20°C.
Catalog No. 50-200-9412