Learn More
Apexbio Technology LLC Thyroid Hormone Receptor Antagonist (1-850),5mg CAS# 251310-57-3
SDP
Supplier:  Apexbio Technology LLC C30065
Encompass_Preferred
Thyroid hormone receptor antagonist (1-850) was screened from a library of more than 250000 compounds as the antagonist of thyroid hormone receptor with the highest affinity. Thyroid Hormone Receptor Antagonist, 1-850 is a cell-permeable hydrazinyl-carboxamide compound which functions as a selective and high-affinity thyroid receptor antagonist with the IC50 value of 1.5 μM in Hela cells [1]. The thyroid hormone receptor (TR) is a type of nuclear receptor that is activated by binding thyroid hormone, which plays a critical role in the regulation of metabolism and heart rate and the development of organisms. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0267