Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thy1/CD90 protein, human (HEK293) | >95.0% | 10 UG
SDP

Catalog No. 5000293254
Encompass_Preferred

Recombinant human Thy1 (CD90) protein expressed in HEK293 cells, provided as a lyophilized, tag-free preparation of the mature Thy1 sequence (Q20-C130). The product is glycosylated and formulated for use as a reference protein or reagent in biochemical and cell biology assays.

  • Human origin, mammalian expression
  • Tag free, glycosylated protein
  • Amino acid range Q20-C130 (mature sequence)
  • Purity >95.0% by reducing SDS-PAGE
  • Endotoxin <1 EU/μg (LAL)
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4
  • Do not reconstitute below 100 μg/mL in ddH2O
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage

Catalog No. 50-002-93254 Supplier Medchemexpress LLC Supplier No. HYP7451810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Thy1 (CD90) protein expressed in HEK293 cells, provided as a lyophilized, tag-free preparation of the mature Thy1 sequence (Q20-C130). The product is glycosylated and formulated for use as a reference protein or reagent in biochemical and cell biology assays.

  • Human origin, mammalian expression
  • Tag free, glycosylated protein
  • Amino acid range Q20-C130 (mature sequence)
  • Purity >95.0% by reducing SDS-PAGE
  • Endotoxin <1 EU/μg (LAL)
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4
  • Do not reconstitute below 100 μg/mL in ddH2O
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.