Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thy1/CD90 protein, human (HEK293) | >=95.0% | 100 UG
SDP

Catalog No. 5000293853
Encompass_Preferred

Recombinant human Thy1/CD90 extracellular domain expressed in HEK293 cells and supplied tag-free. The protein corresponds to human Thy-1 extracellular residues Q20-C130 and is intended for binding assays, assay development, and cell biology research that requires a mammalian-expressed glycoprotein.

  • Recombinant human extracellular domain (Q20-C130).
  • Produced in HEK293 cells for mammalian glycosylation.
  • Tag free to avoid interference in binding studies.
  • Purity ≥95.0% by analytical testing.
  • Available in multiple sizes, including 100 μg.
  • Suitable for binding studies, assay development, and cell biology experiments.

Catalog No. 50-002-93853 Supplier Medchemexpress LLC Supplier No. HYP74518100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Thy1/CD90 extracellular domain expressed in HEK293 cells and supplied tag-free. The protein corresponds to human Thy-1 extracellular residues Q20-C130 and is intended for binding assays, assay development, and cell biology research that requires a mammalian-expressed glycoprotein.

  • Recombinant human extracellular domain (Q20-C130).
  • Produced in HEK293 cells for mammalian glycosylation.
  • Tag free to avoid interference in binding studies.
  • Purity ≥95.0% by analytical testing.
  • Available in multiple sizes, including 100 μg.
  • Suitable for binding studies, assay development, and cell biology experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.