Learn More
Medchemexpress LLC Thy1/CD90 protein, human (HEK293) | >95.0% | 500 UG
SDP
Catalog No. 5000293837
Encompass_Preferred
Recombinant human Thy1/CD90 extracellular-domain protein produced in HEK293 cells, provided as a lyophilized, tag-free powder. The product corresponds to amino acids Q20-C130 of the human Thy1 sequence and is supplied with >95% purity by reducing SDS-PAGE; it is formulated in 20 mM PB, 150 mM NaCl (pH 7.4) prior to lyophilization and intended for research use only.
Catalog No. 50-002-93837 Supplier Medchemexpress LLC Supplier No. HYP74518500UG
Recombinant human Thy1/CD90 extracellular-domain protein produced in HEK293 cells, provided as a lyophilized, tag-free powder. The product corresponds to amino acids Q20-C130 of the human Thy1 sequence and is supplied with >95% purity by reducing SDS-PAGE; it is formulated in 20 mM PB, 150 mM NaCl (pH 7.4) prior to lyophilization and intended for research use only.