Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thy1/CD90 protein, human (HEK293, Fc) | >95.0% | 100 UG
SDP

Catalog No. 5000293976
Encompass_Preferred

Thy1/CD90 protein, human (HEK293, Fc) is a recombinant human Thy-1 ectodomain produced in HEK293 cells and provided as a C-terminal human Fc fusion. Supplied as a filtered PBS solution, it is suitable for research use in binding, cell-interaction, and biochemical assays.

  • Species: human.
  • Source: HEK293 expression system.
  • Tag: C-terminal human Fc fusion.
  • Purity: greater than 95% (reducing SDS-PAGE).
  • Formulation: 0.22 μm filtered PBS, pH 7.4.
  • Molecular weight: approximately 39.3 kDa.
  • Storage and shipping: store at -80°C; ship on dry ice; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-93976 Supplier Medchemexpress LLC Supplier No. HYP74517100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thy1/CD90 protein, human (HEK293, Fc) is a recombinant human Thy-1 ectodomain produced in HEK293 cells and provided as a C-terminal human Fc fusion. Supplied as a filtered PBS solution, it is suitable for research use in binding, cell-interaction, and biochemical assays.

  • Species: human.
  • Source: HEK293 expression system.
  • Tag: C-terminal human Fc fusion.
  • Purity: greater than 95% (reducing SDS-PAGE).
  • Formulation: 0.22 μm filtered PBS, pH 7.4.
  • Molecular weight: approximately 39.3 kDa.
  • Storage and shipping: store at -80°C; ship on dry ice; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.