Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thy1/CD90 protein, human (HEK293, Fc) | >95.0% | 50 UG
SDP

Catalog No. 5000293883
Encompass_Preferred

Recombinant human Thy1 (CD90) extracellular domain fused to a human Fc tag, produced in HEK293 cells and supplied as a sterile PBS solution for in vitro binding and functional assays.

  • Recombinant human Thy-1 ectodomain (Q20-C130) fused to C-terminal human Fc.
  • Produced in HEK293 cells and glycosylated.
  • Supplied as a 0.22 μm filtered solution in PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin less than 1 EU/μg.
  • Molecular weight approximately 39.3 kDa.
  • Stored at -80°C and shipped on dry ice; avoid repeated freeze-thaw.
  • Available in a 50 μg pack size.

Catalog No. 50-002-93883 Supplier Medchemexpress LLC Supplier No. HYP7451750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Thy1 (CD90) extracellular domain fused to a human Fc tag, produced in HEK293 cells and supplied as a sterile PBS solution for in vitro binding and functional assays.

  • Recombinant human Thy-1 ectodomain (Q20-C130) fused to C-terminal human Fc.
  • Produced in HEK293 cells and glycosylated.
  • Supplied as a 0.22 μm filtered solution in PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin less than 1 EU/μg.
  • Molecular weight approximately 39.3 kDa.
  • Stored at -80°C and shipped on dry ice; avoid repeated freeze-thaw.
  • Available in a 50 μg pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.