Learn More
Medchemexpress LLC Thy1/CD90 protein, mouse (HEK293, His) | >95.0% | 10 UG
SDP
Catalog No. 5000293835
Encompass_Preferred
Recombinant, His-tagged full-length mouse Thy1 (CD90) protein expressed in HEK293 cells and supplied as a lyophilized powder. It is intended for biochemical and cell-biology applications that require purified Thy1 protein, including binding studies, antibody validation, and assay development.
  • Full-length mouse Thy1/CD90 expressed in mammalian cells for native folding
  • C-terminal His tag for facile purification and detection
  • High purity (≥95%) as determined by reducing SDS-PAGE
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays
  • Lyophilized formulation for long-term storage and transport
  • Recommended storage at -20°C; aliquot after reconstitution
Catalog No. 50-002-93835 Supplier Medchemexpress LLC Supplier No. HYP7451510UG
Recombinant, His-tagged full-length mouse Thy1 (CD90) protein expressed in HEK293 cells and supplied as a lyophilized powder. It is intended for biochemical and cell-biology applications that require purified Thy1 protein, including binding studies, antibody validation, and assay development.
  • Full-length mouse Thy1/CD90 expressed in mammalian cells for native folding
  • C-terminal His tag for facile purification and detection
  • High purity (≥95%) as determined by reducing SDS-PAGE
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays
  • Lyophilized formulation for long-term storage and transport
  • Recommended storage at -20°C; aliquot after reconstitution