Learn More
Medchemexpress LLC Thy1 (CD90) recombinant human protein | ≥95.0% | 50 UG
SDP
Catalog No. 5000293803
Encompass_Preferred
Thy1 (CD90) recombinant human protein is a tag-free extracellular region produced in HEK293 cells and supplied as a lyophilized powder. It corresponds to sequence Q20-C130 (accession P04216/NP_006279.2) and is intended for biochemical assays, antibody generation, and receptor interaction studies.
  • Lyophilized, tag-free format for flexible reconstitution.
  • Expressed in HEK293 to support human-like glycosylation.
  • Sequence corresponds to Q20-C130, representing the extracellular region.
  • High purity ≥95.0% as determined by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg suitable for sensitive assays.
  • Stable when stored at -20°C; aliquot for long-term storage at -80°C.
  • Available in small research sizes, including 50 μg.
Catalog No. 50-002-93803 Supplier Medchemexpress LLC Supplier No. HYP7451850UG
Thy1 (CD90) recombinant human protein is a tag-free extracellular region produced in HEK293 cells and supplied as a lyophilized powder. It corresponds to sequence Q20-C130 (accession P04216/NP_006279.2) and is intended for biochemical assays, antibody generation, and receptor interaction studies.
  • Lyophilized, tag-free format for flexible reconstitution.
  • Expressed in HEK293 to support human-like glycosylation.
  • Sequence corresponds to Q20-C130, representing the extracellular region.
  • High purity ≥95.0% as determined by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg suitable for sensitive assays.
  • Stable when stored at -20°C; aliquot for long-term storage at -80°C.
  • Available in small research sizes, including 50 μg.