Learn More
Medchemexpress LLC Thymalfasin (Thymosin α1) | 62304-98-7 | 99.3% | 3108.28 | 1 MG
SDP
Catalog No. 5000425698
Encompass_Preferred
Thymalfasin is an immunomodulating agent that can enhance the Th1 immune response. It has demonstrated efficacy in experimental models of immune dysfunction, including infectious diseases like hepatitis and influenza, and cancers such as melanoma and colorectal carcinoma, where it exhibits antitumor effects.
  • Immunomodulating agent: Enhances the Th1 immune response.
  • Effective in experimental models for infectious diseases (e.g., hepatitis, influenza) and various cancers (e.g., melanoma, colorectal carcinoma).
  • Demonstrates antitumor effects in cancer models.
  • High purity: 99.25%.
  • Available in multiple sizes.
Catalog No. 50-004-25698 Supplier Medchemexpress LLC Supplier No. HYP00911MG
Thymalfasin is an immunomodulating agent that can enhance the Th1 immune response. It has demonstrated efficacy in experimental models of immune dysfunction, including infectious diseases like hepatitis and influenza, and cancers such as melanoma and colorectal carcinoma, where it exhibits antitumor effects.
  • Immunomodulating agent: Enhances the Th1 immune response.
  • Effective in experimental models for infectious diseases (e.g., hepatitis, influenza) and various cancers (e.g., melanoma, colorectal carcinoma).
  • Demonstrates antitumor effects in cancer models.
  • High purity: 99.25%.
  • Available in multiple sizes.