Learn More
Medchemexpress LLC Thymalfasin (Thymosin α1) | 62304-98-7 | 99.3% | 3108.28 | 10 MG
SDP
Catalog No. 5000426258
Encompass_Preferred
Thymalfasin is an immunomodulating agent that is capable of enhancing the Thl immune response. It has demonstrated efficacy in various experimental models of immune dysfunction, including infectious diseases like hepatitis (woodchuck) and influenza (mouse), and cancers such as melanoma (mouse) and colorectal carcinoma (rat), where it exhibited antitumor effects.
  • Thymalfasin is an immunomodulating agent.
  • It enhances the Thl immune response.
  • It has shown efficacy in experimental models of immune dysfunction.
  • It has demonstrated antitumor effects in cancer models.
Catalog No. 50-004-26258 Supplier Medchemexpress LLC Supplier No. HYP009110MG
Thymalfasin is an immunomodulating agent that is capable of enhancing the Thl immune response. It has demonstrated efficacy in various experimental models of immune dysfunction, including infectious diseases like hepatitis (woodchuck) and influenza (mouse), and cancers such as melanoma (mouse) and colorectal carcinoma (rat), where it exhibited antitumor effects.
  • Thymalfasin is an immunomodulating agent.
  • It enhances the Thl immune response.
  • It has shown efficacy in experimental models of immune dysfunction.
  • It has demonstrated antitumor effects in cancer models.