Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyme Oil, Red, Spectrum™ Chemical
SDP

Catalog No. 18607398 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
100 kg
Packaging:
Amber Glass Bottle
Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-398 100 kg Drum
18-607-396 100 mL Amber Glass Bottle
18-607-397 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-398 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1047100KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

T1047, 8007-46-3

Spectrum™ Chemical Thyme Oil, Red , an essential oil of thyme, has many applications including as an antiseptic and as an effective agent against certain fungi. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8007-46-3

Specifications

CAS Max % 1
Packaging Drum
Quantity 100 kg
UN Number UN2924
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.