Learn More
CPC Scientific Pyr-Ala-Lys-Ser-Gln-Gly-Gly-Ser-Asn-OH (trifluoroacetate salt) 25MG
Supplier: CPC Scientific IMMO003B
For exerting its anti-inflammatory and immunomodulatory activity, the peptide hormone thymulin, pEAKSQGGSN, requires the presence of Zn(II). Thymulin showed promising results in experimental models of lung diseases as well as potential as an analgesic peptide.ONE-LETTER SEQUENCE: Pyr-AKSQGGSNMOLECULAR FORMULA: C33H54N12O15MOLECULAR WEIGHT:858.86STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [63958-90-7], RESEARCH AREA: ImmunomodulationREFERENCES: J.F.Bach et al., Bull. Inst. Pasteur, 325 (1978); T.L.K.Low and A.L.Goldstein, Springer Semin. Immunopathol., 2, 169 (1979); J.J.E.Haddad et al., Biochem. Biophys. Res. Commun., 274, 500 (2000); S.C.Land and F.Darakshan, Am. J. Physiol. Lung Cell. Mol. Physiol., 286, L473 (2004); M.Dardenne et al., Ann. N. Y. Acad. Sci., 1088, 153 (2006); T.Henriques-Coelho et al., Endocrinology, 149, 4367 (2008); P.C.Reggiani et al., Ann. N. Y. Acad. Sci., 1153, 98 (2009); H.Folch et al., Biol. Res., 43, 291 (2010); M.Santos et al., Expert Opin. T