Learn More
Cayman Chemical ThymIdIn-d4 1mg
SDP
Catalog No. 502911738
Encompass_Preferred
An internal standard for the quantification of thymidine by GC- or LC-MS
Catalog No. 50-291-1738 Supplier Cayman Chemical Supplier No. 363221 MG
Add to Cart
Add to Cart
An internal standard for the quantification of thymidine by GC- or LC-MS