Learn More
Medchemexpress LLC Thymidine-13C | 478511-06-7 | 243.22 | 5 MG
SDP
Catalog No. 5000415136
Encompass_Preferred
Thymidine-13C is the 13C labeled Thymidine. It is a specific precursor of deoxyribonucleic acid and functions as a cell synchronizing agent. Thymidine is also a DNA synthesis inhibitor capable of arresting cells at the G1/S boundary, before DNA replication.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-15136 Supplier Medchemexpress LLC Supplier No. HYN1150S25MG
Thymidine-13C is the 13C labeled Thymidine. It is a specific precursor of deoxyribonucleic acid and functions as a cell synchronizing agent. Thymidine is also a DNA synthesis inhibitor capable of arresting cells at the G1/S boundary, before DNA replication.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.