Learn More
Medchemexpress LLC Thymidine-15N2 | 50-89-5 | 244.22 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN1150S95MG
Encompass_Preferred
Thymidine-15N2 is the 15N labeled Thymidine, a specific precursor of deoxyribonucleic acid. It is used as a cell synchronizing agent and functions as a DNA synthesis inhibitor, capable of arresting cells at the G1/S boundary prior to DNA replication. Stable heavy isotopes, such as 15N, are incorporated into drug molecules to serve as tracers for quantitation during drug development.
  • Used as a tracer and an internal standard for quantitative analysis.
  • Can be analyzed by NMR, GC-MS, or LC-MS techniques.
  • Functions as a DNA synthesis inhibitor.
  • Arrests cells at the G1/S boundary.
  • Suitable for metabolic flux analysis.
  • Applied in various research areas, including cancer, cell cycle, and anti-infection studies.
Catalog No. 50-004-15559