Learn More
Medchemexpress LLC HY-N1150 1g , Thymidine CAS:50-89-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN11501G
Encompass_Preferred
HY-N1150 1g Thymidine CAS: 50-89-5 Thymidine, a specific precursor of deoxyribonucleic acid, is used as a cell synchronizing agent. Thymidine is a DNA synthesis inhibitor that can arrest cell at G1/S boundary, prior to DNA replication. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9580