Learn More
Apexbio Technology LLC Thymidine(Synonyms: Deoxythymidine, Thymine deoxyriboside, dThd, 2'-Deoxythymidine, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, TdR, Thymidin), 25g, CAS: 50-89-5.
SDP
Catalog No. 5000060676
Encompass_Preferred
Thymidine (CAS 50-89-5) chemically known as deoxythymidine is a pyrimidine nucleoside comprising the nitrogenous base thymine linked to a deoxyribose sugar As a fundamental component of DNA thymidine integrates specifically with adenine nucleotides facilitating stable double-helix formation through complementary base pairing Experimentally thymidine incorporation assays are widely employed to monitor DNA synthesis and cell proliferation rates Its utilization in cellular studies allows synchronization of cultured cells at the G1 or early S-phase providing defined conditions to analyze cell cycle dynamics
Catalog No. 50-000-60676 Supplier Apexbio Technology LLC Supplier No. C348625000
Thymidine (CAS 50-89-5) chemically known as deoxythymidine is a pyrimidine nucleoside comprising the nitrogenous base thymine linked to a deoxyribose sugar As a fundamental component of DNA thymidine integrates specifically with adenine nucleotides facilitating stable double-helix formation through complementary base pairing Experimentally thymidine incorporation assays are widely employed to monitor DNA synthesis and cell proliferation rates Its utilization in cellular studies allows synchronization of cultured cells at the G1 or early S-phase providing defined conditions to analyze cell cycle dynamics