Learn More
Medchemexpress LLC Thymidine 3 5 -diph 10mM 1mL | 118675-87-9 | 490.12 | C10H12N2Na4O11P2 | 1 ML
SDP
Catalog No. 5000229384
Encompass_Preferred
Thymidine 3',5'-diphosphate tetrasodium is the tetrasodium salt of deoxythymidine 3',5'-diphosphate provided as a small-volume research reagent for biochemical and molecular biology assays. It is reported to act as an inhibitor of certain nucleases and is used in enzymology and catalytic studies.
  • Provided as a 10 mM solution in 1 mL for ready-to-use assays
  • High purity suitable for research applications
  • Small, convenient volume minimizes waste and supports screening
  • Stable when stored at 4°C and protected from moisture
  • Useful in studies of nuclease inhibition and enzymatic catalysis
Catalog No. 50-002-29384 Supplier Medchemexpress LLC Supplier No. HY115581A10MM 1ML
Thymidine 3',5'-diphosphate tetrasodium is the tetrasodium salt of deoxythymidine 3',5'-diphosphate provided as a small-volume research reagent for biochemical and molecular biology assays. It is reported to act as an inhibitor of certain nucleases and is used in enzymology and catalytic studies.
  • Provided as a 10 mM solution in 1 mL for ready-to-use assays
  • High purity suitable for research applications
  • Small, convenient volume minimizes waste and supports screening
  • Stable when stored at 4°C and protected from moisture
  • Useful in studies of nuclease inhibition and enzymatic catalysis