Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thymidine 3 5 -diph 1mg | 118675-87-9 | 1 MG
SDP

Catalog No. 5000228656
Encompass_Preferred

Thymidine 3',5'-diphosphate tetrasodium is the tetrasodium salt of deoxythymidine diphosphate supplied as a white to off-white solid for research use. It has a molecular formula of C10H12N2Na4O11P2, a molecular weight of 490.12 g/mol, and reported purity of 98.75%. Available in small-mass packages for laboratory experiments.

  • White to off-white solid form, easy to weigh and dissolve
  • High purity (98.75%) suitable for analytical and biochemical work
  • Molecular weight 490.12 g/mol and formula C10H12N2Na4O11P2
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month)
  • Available in small mass packages (1 mg, 5 mg, 10 mg) for research-scale use

Catalog No. 50-002-28656 Supplier Medchemexpress LLC Supplier No. HY115581A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymidine 3',5'-diphosphate tetrasodium is the tetrasodium salt of deoxythymidine diphosphate supplied as a white to off-white solid for research use. It has a molecular formula of C10H12N2Na4O11P2, a molecular weight of 490.12 g/mol, and reported purity of 98.75%. Available in small-mass packages for laboratory experiments.

  • White to off-white solid form, easy to weigh and dissolve
  • High purity (98.75%) suitable for analytical and biochemical work
  • Molecular weight 490.12 g/mol and formula C10H12N2Na4O11P2
  • Stable when stored sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month)
  • Available in small mass packages (1 mg, 5 mg, 10 mg) for research-scale use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.