Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thymidine 3 5 -diph 5mg | 118675-87-9 | 490.12 g/mol | C10H12N2Na4O11P2 | 5 MG
SDP

Catalog No. 5000228638
Encompass_Preferred

Thymidine 3',5'-diphosphate tetrasodium is a nucleotide-based biochemical reagent used in research applications where nucleotide analogs and diphosphate compounds are required. It is supplied as a high-purity tetrasodium salt suitable for in vitro studies and enzymatic assays.

  • High purity: 98.75%.
  • Molecular weight: 490.12 g/mol.
  • Chemical formula: C10H12N2Na4O11P2.
  • Salt form: tetrasodium for improved solubility.
  • Supplied in small laboratory pack sizes, such as a 5 mg vial.

Catalog No. 50-002-28638 Supplier Medchemexpress LLC Supplier No. HY115581A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymidine 3',5'-diphosphate tetrasodium is a nucleotide-based biochemical reagent used in research applications where nucleotide analogs and diphosphate compounds are required. It is supplied as a high-purity tetrasodium salt suitable for in vitro studies and enzymatic assays.

  • High purity: 98.75%.
  • Molecular weight: 490.12 g/mol.
  • Chemical formula: C10H12N2Na4O11P2.
  • Salt form: tetrasodium for improved solubility.
  • Supplied in small laboratory pack sizes, such as a 5 mg vial.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.