Learn More
Medchemexpress LLC Thymidine 5'-monophosphate-13C10,15N2 dilithium | 2483830-49-3 | 99.9% | 345.99 | 13C10H13Li215N2O8P | 10 MG
SDP
Catalog No. 5000285273
Encompass_Preferred
Thymidine 5'-monophosphate-13C10,15N2 dilithium is an isotopically labeled nucleotide provided as a concentrated solution. It carries uniform 13C and 15N labeling on the thymidine moiety and is used as a reference standard and tracer for analytical and structural studies in biochemical research.
  • Isotopically labeled with 13C10 and 15N2.
  • Provided as dilithium salt solution, colorless to light yellow.
  • High purity: 99.9% suitable for analytical applications.
  • Molecular weight 345.99 and CAS 2483830-49-3 for unambiguous identification.
  • Packaged as a concentrated solution for convenient experimental use.
Catalog No. 50-002-85273 Supplier Medchemexpress LLC Supplier No. HYW392933S410MG
Thymidine 5'-monophosphate-13C10,15N2 dilithium is an isotopically labeled nucleotide provided as a concentrated solution. It carries uniform 13C and 15N labeling on the thymidine moiety and is used as a reference standard and tracer for analytical and structural studies in biochemical research.
  • Isotopically labeled with 13C10 and 15N2.
  • Provided as dilithium salt solution, colorless to light yellow.
  • High purity: 99.9% suitable for analytical applications.
  • Molecular weight 345.99 and CAS 2483830-49-3 for unambiguous identification.
  • Packaged as a concentrated solution for convenient experimental use.