Learn More
Apexbio Technology LLC Thymidine-5-monophosphate disodium salt 33430-62-5 100mg
SDP
Catalog No. 5000096143
Encompass_Preferred
Thymidine-5-monophosphate disodium salt (CAS 33430-62-5) is an endogenous nucleotide involved in DNA biosynthesis serving as a metabolic intermediate in the thymidine salvage pathway By influencing the regulation of deoxyribonucleotide pools it modulates DNA replication and repair processes at the cellular level This compound is widely utilized in studies investigating mechanisms of cell proliferation genomic maintenance and DNA damage response Its properties make it a valuable molecular tool in research focused on cancer biology and the development of antiviral therapeutics
Catalog No. 50-000-96143 Supplier Apexbio Technology LLC Supplier No. A8990100
Thymidine-5-monophosphate disodium salt (CAS 33430-62-5) is an endogenous nucleotide involved in DNA biosynthesis serving as a metabolic intermediate in the thymidine salvage pathway By influencing the regulation of deoxyribonucleotide pools it modulates DNA replication and repair processes at the cellular level This compound is widely utilized in studies investigating mechanisms of cell proliferation genomic maintenance and DNA damage response Its properties make it a valuable molecular tool in research focused on cancer biology and the development of antiviral therapeutics