Learn More
Medchemexpress LLC Thymidine 5'-monophosphate p-nitrophenyl ester sodium | 98179-10-3 | 99.9% | C16H17N3NaO10P | 10MG
SDP
Catalog No. 5000259658
Encompass_Preferred
Thymidine 5′-monophosphate p-nitrophenyl ester sodium is a chemically modified thymidine 5′-phosphate derivative provided as the sodium salt for biochemical research. It penetrates mammalian cells and is reported to convert intracellularly to thymidine 5′-phosphate (TMP), and has been investigated as a potential antitumor prodrug. Analytical documentation is provided for research applications.
  • Cell-permeable thymidine phosphate derivative suitable for biochemical studies.
  • Reported intracellular conversion to thymidine 5′-phosphate (TMP).
  • Provided as a sodium salt for aqueous compatibility.
  • Supplied with purity documentation including COA, HNMR, and LCMS.
  • Available in small research-scale quantities.
Catalog No. 50-002-59658 Supplier Medchemexpress LLC Supplier No. HY13780710MG
Add to Cart
Add to Cart
Thymidine 5′-monophosphate p-nitrophenyl ester sodium is a chemically modified thymidine 5′-phosphate derivative provided as the sodium salt for biochemical research. It penetrates mammalian cells and is reported to convert intracellularly to thymidine 5′-phosphate (TMP), and has been investigated as a potential antitumor prodrug. Analytical documentation is provided for research applications.
  • Cell-permeable thymidine phosphate derivative suitable for biochemical studies.
  • Reported intracellular conversion to thymidine 5′-phosphate (TMP).
  • Provided as a sodium salt for aqueous compatibility.
  • Supplied with purity documentation including COA, HNMR, and LCMS.
  • Available in small research-scale quantities.