Learn More
Medchemexpress LLC Thymidine 5′-monophosphate p-nitrophenyl ester sodium | 98179-10-3 | MFCD00036763 | 99.9% | 465.28 | C16H17N3NaO10P | 5 MG
SDP
Catalog No. 5000223481
Encompass_Preferred
Thymidine 5′-monophosphate p-nitrophenyl ester sodium is a synthetic nucleotide derivative supplied as the sodium salt for research use. It functions as a chromogenic substrate for enzymatic and analytical assays, facilitating colorimetric detection of phosphatase and nucleotidase activity. Supplied as a solid with high purity for laboratory applications.
  • High purity suitable for analytical and biochemical assays.
  • Sodium salt form provided as a solid for ease of handling and storage.
  • Chromogenic p-nitrophenyl ester enables colorimetric detection.
  • Available in small package sizes for research-scale experiments.
  • Useful as a substrate to assess nucleotidase and phosphatase activity.
Catalog No. 50-002-23481 Supplier Medchemexpress LLC Supplier No. HY1378075MG
Thymidine 5′-monophosphate p-nitrophenyl ester sodium is a synthetic nucleotide derivative supplied as the sodium salt for research use. It functions as a chromogenic substrate for enzymatic and analytical assays, facilitating colorimetric detection of phosphatase and nucleotidase activity. Supplied as a solid with high purity for laboratory applications.
  • High purity suitable for analytical and biochemical assays.
  • Sodium salt form provided as a solid for ease of handling and storage.
  • Chromogenic p-nitrophenyl ester enables colorimetric detection.
  • Available in small package sizes for research-scale experiments.
  • Useful as a substrate to assess nucleotidase and phosphatase activity.