Learn More
Medchemexpress LLC HY-N1150 500mg Medchemexpress, Thymidine CAS:50-89-5 Purity:>98%
SDP
Catalog No. 501970326
Encompass_Preferred
Medchemexpress, HY-N1150 500mg Thymidine CAS:50-89-5 Thymidine, a specific precursor of deoxyribonucleic acid, is used as a cell synchronizing agent. Thymidine is a DNA synthesis inhibitor that can arrest cell at G1/S boundary, prior to DNA replication[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0326 Supplier Medchemexpress LLC Supplier No. HYN1150500MG
Medchemexpress, HY-N1150 500mg Thymidine CAS:50-89-5 Thymidine, a specific precursor of deoxyribonucleic acid, is used as a cell synchronizing agent. Thymidine is a DNA synthesis inhibitor that can arrest cell at G1/S boundary, prior to DNA replication[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.