Learn More
Medchemexpress LLC Thymidine-d3 | 74848-84-3 | MFCD01317525 | 99.8% | 245.25 | C10H11D3N2O5 | 10 MG
SDP
Catalog No. 5000296164
Encompass_Preferred
Thymidine-d3 is the deuterium-labeled analogue of thymidine used as a DNA synthesis inhibitor and cell-synchronizing agent. It is commonly used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, and to arrest cells at the G1/S boundary in cell cycle studies.
  • High chemical and isotopic purity (99.8%).
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Effective for cell synchronization by arresting cells at the G1/S boundary.
  • Solid powder form with recommended cold storage for stability.
  • Available in small research pack sizes including 10 mg for analytical work.
Catalog No. 50-002-96164 Supplier Medchemexpress LLC Supplier No. HYN1150S10MG
Thymidine-d3 is the deuterium-labeled analogue of thymidine used as a DNA synthesis inhibitor and cell-synchronizing agent. It is commonly used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, and to arrest cells at the G1/S boundary in cell cycle studies.
  • High chemical and isotopic purity (99.8%).
  • Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Effective for cell synchronization by arresting cells at the G1/S boundary.
  • Solid powder form with recommended cold storage for stability.
  • Available in small research pack sizes including 10 mg for analytical work.