Learn More
Medchemexpress LLC Thymidine-d4 | 347841-67-2 | 246.25 | 1 MG
SDP
Catalog No. 5000415064
Encompass_Preferred
Thymidine-d4 is a deuterium-labeled version of Thymidine, which acts as a specific precursor for deoxyribonucleic acid. It is employed as a cell synchronizing agent and functions as a DNA synthesis inhibitor, halting cell progression at the G1/S boundary before DNA replication. Stable heavy isotopes like deuterium are integrated into drug molecules primarily as tracers for quantitative analysis during drug development, with deuteration specifically considered for its potential to alter the pharmacokinetic and metabolic profiles of drugs.
  • Is the deuterium labeled Thymidine.
  • Is a specific precursor of deoxyribonucleic acid.
  • Used as a cell synchronizing agent.
  • Functions as a DNA synthesis inhibitor.
  • Arrests cell at G1/S boundary, prior to DNA replication.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15064 Supplier Medchemexpress LLC Supplier No. HYN1150S11MG
Thymidine-d4 is a deuterium-labeled version of Thymidine, which acts as a specific precursor for deoxyribonucleic acid. It is employed as a cell synchronizing agent and functions as a DNA synthesis inhibitor, halting cell progression at the G1/S boundary before DNA replication. Stable heavy isotopes like deuterium are integrated into drug molecules primarily as tracers for quantitative analysis during drug development, with deuteration specifically considered for its potential to alter the pharmacokinetic and metabolic profiles of drugs.
  • Is the deuterium labeled Thymidine.
  • Is a specific precursor of deoxyribonucleic acid.
  • Used as a cell synchronizing agent.
  • Functions as a DNA synthesis inhibitor.
  • Arrests cell at G1/S boundary, prior to DNA replication.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.