Learn More
Medchemexpress LLC Thymidine-d4 | 347841-67-2 | 246.25 | 5 MG
SDP
Catalog No. 5000416307
Encompass_Preferred
Thymidine-d4 is deuterium-labeled Thymidine. Thymidine is a specific precursor of deoxyribonucleic acid and functions as a cell synchronizing agent. It also acts as a DNA synthesis inhibitor, capable of arresting cells at the G1/S boundary prior to DNA replication. This compound is useful in various analytical and research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs
  • Stable heavy isotopes can serve as tracers for quantitation during drug development
Catalog No. 50-004-16307 Supplier Medchemexpress LLC Supplier No. HYN1150S15MG
Thymidine-d4 is deuterium-labeled Thymidine. Thymidine is a specific precursor of deoxyribonucleic acid and functions as a cell synchronizing agent. It also acts as a DNA synthesis inhibitor, capable of arresting cells at the G1/S boundary prior to DNA replication. This compound is useful in various analytical and research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs
  • Stable heavy isotopes can serve as tracers for quantitation during drug development