Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidine, [Methyl-3H], MP Biomedicals™

Catalog No. MP012406601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1mCi; 37MBq
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP012406601 1mCi; 37MBq
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP012406601 Supplier MP Biomedicals, Inc Supplier No. 012406601

Specifications

Physical Form Sterile Aqueous Solution
Quantity 1 mCi
Molecular Formula C10H14N2O5
Formula Weight 242.2
Chemical Name or Material Thymidine, [Methyl-3H]
Grade HPLC analyzed for purity
Recommended Storage Store at 2-8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.