Learn More
Revvity Health Sciences Inc Thymidine, [Methyl-3H]-, Specific Activity: 70-90Ci (2.59-3.33TBq)/mmol,>97%, 5mCi (185MBq)
Catalog No. 502810773
Encompass
Thymidine, [Methyl-3H]-, Specific Activity: 70-90Ci (2.59-3.33TBq)/mmol,>97%, 5mCi (185MBq)
Catalog No. 50-281-0773 Supplier Revvity Health Sciences Inc Supplier No. NET027Z005MC
Thymidine, [Methyl-3H]-, Specific Activity: 70-90Ci (2.59-3.33TBq)/mmol,>97%, 5mCi (185MBq)