Learn More
Revvity Health Sciences Inc Thymidine, [Methyl-3H]-, Specific Activity: 70-90Ci (2.59-3.33TBq)/mmol,>97%, 250µCi (9.25MBq)
Catalog No. 502810536
Encompass
Thymidine, [Methyl-3H]-, Specific Activity: 70-90Ci (2.59-3.33TBq)/mmol,>97%, 250µCi (9.25MBq)
Catalog No. 50-281-0536 Supplier Revvity Health Sciences Inc Supplier No. NET027Z250UC
Thymidine, [Methyl-3H]-, Specific Activity: 70-90Ci (2.59-3.33TBq)/mmol,>97%, 250µCi (9.25MBq)