Learn More
Abcam Thymidine, Pyrimidine deo x ynucleoside, 10G
Catalog No. 502706092
Encompass_Preferred
MW 242.23 Da, Purity >98%. Pyrimidine deoxynucleoside. Potent thymidine kinase substrate (IC₅₀ = 16 nM). Pairs with deoxyadenosine in double-stranded DNA. Synchronizes cells in S phase in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6092 Supplier Abcam Supplier No. AB14371910G
MW 242.23 Da, Purity >98%. Pyrimidine deoxynucleoside. Potent thymidine kinase substrate (IC₅₀ = 16 nM). Pairs with deoxyadenosine in double-stranded DNA. Synchronizes cells in S phase in vivo. .
The product is subject to the following: Abcam Restricted Use Statement