Learn More
Cayman Chemical Thymine-13C5,15N2, Purity: ≥98%, CAS: 2483830-13-1, 1 mg
SDP
Catalog No. 502944620
Encompass_Preferred
An internal standard for the quantification of thymine by GC- or LC-MS
Catalog No. 50-294-4620 Supplier Cayman Chemical Supplier No. 411721 MG
An internal standard for the quantification of thymine by GC- or LC-MS