Learn More
Cayman Chemical ThymIn-13C5 15N2 500ug
SDP
Catalog No. 502943530
Encompass_Preferred
An internal standard for the quantification of thymine by GC- or LC-MS
Catalog No. 50-294-3530 Supplier Cayman Chemical Supplier No. 41172500 UG
An internal standard for the quantification of thymine by GC- or LC-MS