Learn More
Medchemexpress LLC Thymine | MFCD00006026 | 99.9% | 129.09 g/mol | C5H6N2O2 | 1mg
SDP
Catalog No. 5000269037
Encompass_Preferred
Thymine-15N2 13C is the 13C and 15N labeled Thymine (HY-W010450) Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-Fluorouracil (HY-90006) in cancer treatment with a Km of 2 3 M[1]
Catalog No. 50-002-69037 Supplier Medchemexpress LLC Supplier No. HYW7472141MG
Thymine-15N2 13C is the 13C and 15N labeled Thymine (HY-W010450) Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-Fluorouracil (HY-90006) in cancer treatment with a Km of 2 3 M[1]