Learn More
Medchemexpress LLC Thymine 1-β-D-arabinofuranoside | 605-23-2 | 98.0% | 258.23 | 25 MG
SDP
Catalog No. 5000352143
Encompass_Preferred
Thymine 1-β-D-arabinofuranoside is a purine nucleoside analogue. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms include inhibition of DNA synthesis and induction of apoptosis.
  • Purine nucleoside analogue
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Solid appearance
  • Off-white to light yellow color
  • Melting/freezing point: 246-247°C
  • Recommended storage: 4°C, stored under nitrogen
Catalog No. 50-003-52143 Supplier Medchemexpress LLC Supplier No. 25MGHY137848
Thymine 1-β-D-arabinofuranoside is a purine nucleoside analogue. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms include inhibition of DNA synthesis and induction of apoptosis.
  • Purine nucleoside analogue
  • Exhibits broad antitumor activity
  • Targets indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • Solid appearance
  • Off-white to light yellow color
  • Melting/freezing point: 246-247°C
  • Recommended storage: 4°C, stored under nitrogen