Learn More
Medchemexpress LLC HY-W010450 1g Medchemexpress, Thymine CAS:65-71-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0104501G
Encompass_Preferred
Medchemexpress, HY-W010450 1g Thymine CAS:65-71-4 Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment, with a K m of 2.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4538