Learn More
Alkali Scientific Thymine, 25 G
SDP
Supplier:  Alkali Scientific HYM001CP
Encompass_Preferred
Thymine is a nucleobase essential for DNA structure, acting as one of the building blocks of genetic material. This chemical is widely used in laboratory applications involving nucleic acid synthesis and analysis. Thymine, a pyrimidine base, pairs specifically with adenine in double-stranded DNA through hydrogen bonds, playing a critical role in the genetic code’s integrity. It is used in biochemical research, including studies on DNA replication, repair, and transcription. The 25g format is convenient for laboratory-scale experiments, including DNA synthesis, sequencing, and various enzymatic reactions. Thymine is commonly utilized in the production of synthetic oligonucleotides and other biochemical reagents.
Catalog No. 50-006-59640