Learn More
Medchemexpress LLC Thymine-d4-1 | 156054-85-2 | 99.47% | 130.14 | 50 MG
SDP
Catalog No. 5000367082
Encompass_Preferred
Thymine-d4-1 is the deuterium labeled Thymine. Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM. Stable heavy isotopes have been incorporated into drug molecules for use as tracers and for quantitation during drug development. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Suitable for research use only
Catalog No. 50-003-67082 Supplier Medchemexpress LLC Supplier No. HYW010450S450MG
Thymine-d4-1 is the deuterium labeled Thymine. Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM. Stable heavy isotopes have been incorporated into drug molecules for use as tracers and for quantitation during drug development. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Suitable for research use only